Order Entry
United Kingdom
ContactUsLinkComponent
Anti-RGMB/Repulsive Guidance Molecule B Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Anti-RGMB/Repulsive Guidance Molecule B Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Catalog # BOSSBS-11474R-A750
Supplier:  Bioss
Anti-RGMB/Repulsive Guidance Molecule B Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Catalog # BOSSBS-11474R-A750
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    RGMB/Repulsive Guidance Molecule B
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 750
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Epitope:
    58-160/437
  • Form:
    Liquid
  • Antigen synonyms:
    Rgmb|Dragon|RGM b|RGM domain family member B|RGMB_HUMAN|Repulsive Guidance Molecule B|DRG11-responsive axonal guidance and outgrowth of neurite
  • Modification:
    Unmodified
  • Storage buffer:
    Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
  • Molecular weight:
    40 kDa
  • Storage temperature:
    Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
  • Concentration:
    1 µg/µl
  • Shipping temperature:
    4 °C
  • Immunogen:
    KLH conjugated synthetic peptide derived from human Repulsive Guidance Molecule B
  • Tested applications:
    ICC
  • Purification:
    Purified by Protein A
  • Pack type:
    Vial
  • Pk:
    100 µl

Specifications

About this item

The repulsive guidance molecule (RGM) family of proteins are important in the guidance of growth cones of developing neurons. They are repulsive for a group of axons, those from the temporal half of the retina. RGM have been implicated in both axonal guidance and neural tube closure but as opposed to for ephrins, semaphorins, netrins and slits, no receptor mechanism for RGM activation has been defined. Dorsal root ganglion axons do not respond to RGM but neogenin (a netrin-binding protein which can function as an RGM receptor) expression can spur RGM responsiveness. The RGM proteins are attached to the membrane by a GPI-anchor. Two members of this family, RGMa and RGMb, are expressed in the nervous system. RGMc, also known as Hemojuvelin, is a part of the Signalling pathway activating hepcidin and works together with hepcidin to restrict iron absorption in the gut. Defects in the gene encoding for RGMc causes the autosomal recessive disorder juvenile hemochromatosis (JH).

Type: Primary
Antigen: RGMB/Repulsive Guidance Molecule B
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 58-160/437
Host: Rabbit
Isotype: IgG
Reactivity: