Anti-Collagen alpha-1(II) chain Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Catalog # BOSSBS-10589R-A750
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Collagen alpha-1(II) chain
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 750
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Rabbit,Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Epitope:231-330/1487
- Form:Liquid
- Gene ID:1280
- Antigen synonyms:COL2A1|Alpha-1 type II collagen|SEDC|AOM|STL1|Collagen alpha-1(II) chain|ANFH|COL11A3
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:117 kDa
- Storage temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human Collagen alpha-1(II) chain
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
Type II collagen is specific for cartilaginous tissues. It is essential for the normal embryonic development of the skeleton, for linear growth and for the ability of cartilage to resist compressive forces.
Type: Primary
Antigen: Collagen alpha-1(II) chain
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 231-330/1487
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Rabbit