Anti-GnRH associated peptide I Rabbit Polyclonal Antibody (Alexa Fluor® 750)
BOSSBS-10370R-A750
: Bioss
- Antibody type:Primary
- Antigen name:GnRH associated peptide I
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 750
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:
- Environmentally Preferable:
- Epitope:21-80/92
- Form:Liquid
- Gene ID:2796
- Antigen synonyms:GNRH|Luliberin I|LH-RH I|Luteinizing hormone-releasing hormone I|GON1_HUMAN|LHRH|GRH|progonadoliberin-1 isoform 2 preproprotein|GnRH-associated peptide I|GnRH-I|Gonadotropin-releasing hormone I|GNRH1|Progonadoliberin-1|progonadoliberin-1 isoform 1 precursor|Progonadoliberin I|GnRH-associated peptide 1
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:8/6 kDa
- Storage temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human peptide I
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
The protein encoded by this gene is secreted and then cleaved to form the 10 aa luteinizing hormone-releasing hormone (LHRH, also known as gonadoliberin-1), and prolactin release-inhibiting factor (also known as GnRH-associated peptide 1). LHRH stimulates the release of luteinizing and follicle stimulating hormones, which are important for reproduction. Mutation in this gene are associated with hypogonadotropic hypogonadism. Alternatively spliced transcript variants have been described for this gene.
Type: Primary
Antigen: GnRH associated peptide I
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 21-80/92
Host: Rabbit
Isotype: IgG
Reactivity: