Anti-SCNN1D Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Catalog # BOSSBS-10141R-A750
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:SCNN1D
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 750
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Rat
- Environmentally Preferable:
- Epitope:451-550/638
- Form:Liquid
- Gene ID:6339
- Antigen synonyms:Amiloride-sensitive sodium channel subunit delta|ENaCd|Delta-NaCH|Nonvoltage-gated sodium channel 1 subunit delta|Delta-ENaC|SCNN1D|SCNED|ENaCdelta|dNaCh|Epithelial Na(+) channel subunit delta
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:70 kDa
- Storage temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human SCNN1D
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
Sodium permeable non-voltage-sensitive ion channel inhibited by the diuretic amiloride. Mediates the electrodiffusion of the luminal sodium (and water, which follows osmotically) through the apical membrane of epithelial cells. Controls the reabsorption of sodium in kidney, colon, lung and sweat glands. Also plays a role in taste perception.
Type: Primary
Antigen: SCNN1D
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 451-550/638
Host: Rabbit
Isotype: IgG
Reactivity: Rat