Anti-H-1 beta Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Catalog # BOSSBS-1407R-A555
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:H-1 beta
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 555
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Environmentally Preferable:
- Epitope:681-789/789
- Form:Liquid
- Gene ID:405
- Antigen synonyms:HIF-1-beta|HIF1BETA|HIF-1beta|Hypoxia-inducible factor 1-beta|Class E basic helix-loop-helix protein 2|bHLHe2|ARNT protein|TANGO|HIF1B|Dioxin receptor, nuclear translocator|ARNT|HIF1-beta|Aryl hydrocarbon receptor nuclear translocator
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:87 kDa
- Storage temperature:Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human HIF-1 Beta
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
Required for activity of the Ah (dioxin) receptor. This protein is required for the ligand-binding subunit to translocate from the cytosol to the nucleus after ligand binding. The complex then initiates transcription of genes involved in the activation of PAH procarcinogens. The heterodimer with HIF1A or EPAS1/HIF2A functions as a transcriptional regulator of the adaptive response to hypoxia.
Type: Primary
Antigen: H-1 beta
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 555
Public Immunogen Range: 681-789/789
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse