Anti-CSP Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Catalog # BOSSBS-0848R-A750
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:CSP
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 750
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Other
- Environmentally Preferable:
- Epitope:101-200/367
- Form:Liquid
- Antigen synonyms:circumsporozoite protein Plasmodium yoelii|Circumsporozoite protein
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:37 kDa
- Storage temperature:Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from Plasmodium yoelii CSP
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
The circumsporozoite protein is the immunodominant surface antigen on the sporozoite.
Type: Primary
Antigen: CSP
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 101-200/367
Host: Rabbit
Isotype: IgG
Reactivity: Others