Anti-ADRB1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Catalog # BOSSBS-0498R-A680
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:ADRB1
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 680
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Epitope:181-250/477
- Form:Liquid
- Gene ID:153
- Antigen synonyms:RHR|BETA1AR|ADRB1|Beta-1 adrenoceptor|B1AR|Beta-1 adrenergic receptor|ADRB1R|Beta-1 adrenoreceptor
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:52 kDa
- Storage temperature:Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human ADRB1
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
Beta-adrenergic receptors mediate the catecholamine-induced activation of adenylate cyclase through the action of G proteins. This receptor binds epinephrine and norepinephrine with approximately equal affinity. Mediates Ras activation through G(s)-alpha- and cAMP-mediated signaling.
Type: Primary
Antigen: ADRB1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 181-250/477
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat