Anti-Ghrelin Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Catalog # BOSSBS-0467R-A750
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Ghrelin
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 750
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat
- Western blot:Yes
- Environmentally Preferable:
- Epitope:31-117/117
- Form:Liquid
- Gene ID:51738
- Antigen synonyms:UNQ524/PRO1066|Growth hormone-releasing peptide|MTLRP|Protein M46|Motilin-related peptide|Appetite-regulating hormone|GHRL|Growth hormone secretagogue
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:13 kDa
- Storage temperature:Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human Ghrelin
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
Ghrelin is the ligand for growth hormone secretagogue receptor type 1 (GHSR). Induces the release of growth hormone from the pituitary. Has an appetite-stimulating effect, induces adiposity and stimulates gastric acid secretion. Involved in growth regulation. Obestatin may be the ligand for GPR39. May have an appetite-reducing effect resulting in decreased food intake. May reduce gastric emptying activity and jejunal motility (By similarity).
Type: Primary
Antigen: Ghrelin
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 31-117/117
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat