Anti-NT5C2 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Catalog # BOSSBS-0372R-A750
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:NT5C2
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 750
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Rat
- Western blot:Yes
- Environmentally Preferable:
- Epitope:1-100/561
- Form:Liquid
- Gene ID:22978
- Antigen synonyms:NT5CP|PNT5|SPG45|GMP|NT5B|SPG65|NT5C2|Cytosolic purine 5'-nucleotidase|cN-II|Cytosolic 5'-nucleotidase II
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:65 kDa
- Storage temperature:Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human NT5C2
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
May have a critical role in the maintenance of a constant composition of intracellular purine/pyrimidine nucleotides in cooperation with other nucleotidases. Preferentially hydrolyses inosine 5'-monophosphate (IMP) and other purine nucleotides.
Type: Primary
Antigen: NT5C2
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range:1-100/561
Host: Rabbit
Isotype: IgG
Reactivity: Rat