Order Entry
United Kingdom
ContactUsLinkComponent
Anti-HMGCS1 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Anti-HMGCS1 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
  BOSSBS-5069R-A750
 :  Bioss
Anti-HMGCS1 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
  BOSSBS-5069R-A750
 :  Bioss

 

  • Antibody type:
    Primary
  • Antigen name:
    HMGCS1
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 750
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Mouse
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Epitope:
    41-150/520
  • Form:
    Liquid
  • Gene ID:
    266977
  • Antigen synonyms:
    HMCS1_HUMAN|HMGCS|3 hydroxy 3 methylglutaryl Coenzyme A synthase 1 soluble|MGC90332: EC 2.3.3.10|HMG CoA synthase|Hydroxymethylglutaryl CoA synthase, cytoplasmic
  • Modification:
    Unmodified
  • Storage buffer:
    Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
  • Molecular weight:
    57 kDa
  • Storage temperature:
    Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
  • Concentration:
    1 µg/µl
  • Shipping temperature:
    4 °C
  • Immunogen:
    KLH conjugated synthetic peptide derived from human HMGCS1
  • Tested applications:
    ICC
  • Purification:
    Purified by Protein A
  • Pack type:
    Vial
  • Pk:
    100 µl

 

 

This enzyme condenses acetyl CoA with acetoacetyl CoA to form HMG CoA, which is the substrate for HMG CoA reductase.

Type: Primary
Antigen: HMGCS1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 41-150/520
Host: Rabbit
Isotype: IgG
Reactivity: Mouse