Anti-FPRL1/Lipoxin A4 receptor Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Catalog # BOSSBS-3654R-A750
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:FPRL1/Lipoxin A4 receptor
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 750
- Flow cytometry:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Epitope:51-150/351
- Form:Liquid
- Gene ID:2358
- Antigen synonyms:FPRH1|FPRH2|FMLPX|FMLP-R-I|FPRL1|FMLP-related receptor I|FPR2A|FMLP-R-II|HM63|LXA4R|N-formyl peptide receptor 2|FPR2|Formyl peptide receptor-like 1|RFP|Lipoxin A4 receptor|ALXR|LXA4 receptor
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:39 kDa
- Storage temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human FPRL1
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
Low affinity receptor for N-formyl-methionyl peptides, which are powerful neutrophils chemotactic factors. Binding of FMLP to the receptor causes activation of neutrophils. This response is mediated via a G-protein that activates a phosphatidylinositol-calcium second messenger system. The activation of LXA4R could result in an anti-inflammatory outcome counteracting the actions of proinflammatory signals such as LTB4 (leukotriene B4).
Type: Primary
Antigen: FPRL1/Lipoxin A4 receptor
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 51-150/351
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat