- Antibody type:Primary
- Antigen name:HCP5
- Clonality:Polyclonal
- Conjugation:Cy7®
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Epitope:1-80/132
- Form:Liquid
- Antigen synonyms:HCP5_HUMAN|HLA complex protein P5|HLA class I histocompatibility antigen protein P5|Protein P5 1
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:14 kDa
- Storage temperature:Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human HCP5
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
HCP5 is a region present on chromosome 6p21.3 that is characterised by multiple duplicated gene families. HCP5 (HLA class I histocompatibility antigen protein P5), also known as P5-1, is a 132 amino acid protein that is encoded by a gene mapping to human chromosome 6p21.33. Localising within the MHC class I region, HCP5 is not structurally related to other MHC class I genes, but does have high sequence similarity with HERV-L and HERV-16. HCP5 also has high sequence homology to retroviral Pol genes, making it a possible candidate for interaction with HIV-1 through an antisense mechanism that prevents retrovirus transcription. Single-nucleotide polymorphisms (SNPs) to the region of the HCP5 gene that corresponds with HLA-B has been linked to a lower HIV-1 viral set point. HCP5 is highly expressed in lymphoid tissues, spleen and activated lymphocytes, as well as B-cell and natural killer (NK) cell lines.
Type: Primary
Antigen: HCP5
Clonality: Polyclonal
Clone:
Conjugation: Cy7
Public Immunogen Range: 1-80/132
Host: Rabbit
Isotype: IgG
Reactivity: