Anti-GPR10 Rabbit Polyclonal Antibody (ALEXA FLUOR® 555)
Catalog # BOSSBS-15354R-A555
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:GPR10
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 555
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Epitope:151-250/370
- Form:Liquid
- Antigen synonyms:PrRP receptor|PRLHR|Prolactin-releasing peptide receptor|PrRPR|GPR10|hGR3|PRLHR_HUMAN|MGC126539|MGC126541|GPCR 10|G-protein coupled receptor 10|GR3
- Modification:Unmodified
- Storage buffer:Aqueous buffered solution containing 0,01 M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol
- Molecular weight:41 kDa
- Storage temperature:Store at –20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic peptide derived from human GPR10 protein
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
GPR10.
Type: Primary
Antigen: GPR10
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 555
Public Immunogen Range: 151-250/370
Host: Rabbit
Isotype: IgG
Reactivity: