Anti-FABP4 Tyr20 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Catalog # BOSSBS-13063R-A680
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:FABP4 Tyr20
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 680
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Form:Liquid
- Gene ID:2167
- Antigen synonyms:adipocyte|422/aP2|FABP4_HUMAN|Adipocyte lipid binding protein|Fatty acid binding protein 4|ALBPv ALBP/Ap2|p-FABP4 Y20|Protein 422|Myelin P2 protein homolog|FABP4 phospho Y20|A-FABP|Fatty acid binding protein 4 adipocyte|Fatty acid-binding protein 4|Fatty acid-binding protein|FABP4|Lbpl|P15v P2 adipocyte protein|FABP|3T3-L1 lipid-binding protein|Fatty acid binding protein adipocyte|p-FABP4 Tyr20|Adipocyte lipid-binding protein|Adipocyte protein AP2|AFABP|A FABP|aP2|FABP4 phospho Tyr20|Adipocyte-type fatty acid-binding protein
- Modification:Phosphorylation (Tyr20)
- Storage buffer:Aqueous buffered solution containing 0,01M TBS (pH 7,4) with 1% BSA, 0,03% Proclin300 and 50% Glycerol.
- Molecular weight:14.6 kDa
- Storage temperature:Store at −20 °C. Aliquot into multiple vials to avoid repeated freeze-thaw cycles.
- Concentration:1 µg/µl
- Shipping temperature:4 °C
- Immunogen:KLH conjugated synthetic phosphopeptide derived from human FABP4 around the phosphorylation site of Tyr20
- Tested applications:ICC
- Purification:Purified by Protein A
- Pack type:Vial
- Pk:100 µl
Specifications
About this item
FABP4 encodes the fatty acid binding protein found in adipocytes. Fatty acid binding proteins are a family of small, highly conserved, cytoplasmic proteins that bind long-chain fatty acids and other hydrophobic ligands. It is thought that FABPs roles include fatty acid uptake, transport, and metabolism. [provided by RefSeq].
Type: Primary
Antigen: FABP4 Tyr20
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range:
Host: Rabbit
Isotype: IgG
Reactivity: