Order Entry
United Kingdom
ContactUsLinkComponent
Human Type AB Serum, MP Biomedicals
  ICNA092930149
undefined
Human Type AB Serum, MP Biomedicals
  ICNA092930149
  • Description:
    Human serum, pooled
  • Environmentally Preferable:
  • Pk:
    100 mL
Human Type AB Serum is used as a growth supplement for cell culture media and in other research applications.
  • Collected from Type AB human blood
  • Liquid media
  • Storage at —20 °C
It has been proven to grow many human cell lines at a faster rate and with a smaller percentage of serum than mixed blood group serum.