Order Entry
United Kingdom
ContactUsLinkComponent
GCQ CleanType® Medical Silicone Keyboard with Illumination
GCQ CleanType® Medical Silicone Keyboard with Illumination
Catalog # GETTKG24229
Supplier:  GETT GERATETECHNIK
undefined
GCQ CleanType® Medical Silicone Keyboard with Illumination
Catalog # GETTKG24229
Supplier:  GETT GERATETECHNIK
Specifications
  • Description:
    TKG-110-GCQ-MED, IP 68, USB, backlit, white, DE layout
  • W×D×H:
    380×130×13 mm
  • Environmentally Preferable:
  • Pk:
    1
Specifications
About this item
The CleanType® Medical keyboard is specially manufactured for medical environments. With its versatility, it can be used in almost all areas of hospitals, clinics, and medical practices.
  • IP 68 rated medical keyboard
  • Fully washable and disinfectable
  • Water-resistant silicone USB protection cap
  • IEC 60601-1-2 certified
  • For use in various areas: hospitals, offices, laboratories, production areas
The CleanType® Medical keyboard is completely waterproof, can be disinfected, and features integrated backlighting. The keyboard is a product of the GCQ series from GETT.
Certifications: CE, FCC, REACH.
Packaging: Keyboards are packed and delivered in environmentally friendly brown cardboard.