Smiths Medical® Luer Fittings on PVC Flexible Tubing, Straight
Get a seamless transition from source to system.
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimise downtime
- Regulatory approvals to match your unique application
- Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
This high pressure flexible PVC tubing has luer fittings at each end and comes in various lengths. Can be sterilized by ETO.
- Environmentally Preferable:
- Catalog No:
- MFLX30526-14BLK
- MFLX30526-16BLK
- Connection type (port 1):
- Female luer
- Female luer
- Connection type (port 2):
- Male luer lock
- Male luer lock
- No. of ports:
- 2
- 2
- Warranty:
- Out of box
- Out of box
Frequently Bought Together