Order Entry
United Kingdom
ContactUsLinkComponent
Heel grounder, Wescorp
Heel grounder, Wescorp
Catalog # 115-3062
Supplier:  DESCO
Heel grounder, Wescorp
Catalog # 115-3062
Supplier:  DESCO
Specifications
  • Environmentally Preferable:
  • Pk:
    1
Specifications
About this item
Wescorp heel ground with one Megohm resistor are designed to satisfy Footwear requirements of IEC 61340-5-1.
  • Wear resistant rubber
  • Provides contact area to floor
  • Non-marking interior
  • Will not mark shoes
  • Snap-loc fastening assembly
  • Genuine non-fraying Hook and Loop material - adjustable and comfortable
  • 30,5 cm long, 8 strand nylon polyester grounding tab
  • Does not particulate; superior body to ground contact
  • Rg: 1×10⁶ < ×10⁷ Ω per IEC 61340-5-1 Clause A.2
Per IEC 61340-5-2 paragraph 5.2.8, footwear should be used 'where personnel are mobile, in storing areas, operating large machinery, etc., it may be impractical or dangerous to have long cords attached to them.'