About this item
The CleanType® Easy Basic is a full layout, high-quality plastic keyboard that is easy to clean thanks to its water and dust resistance.
- Washable plastic keyboard
- Waterproof and dustproof
- Available with white or black finish
- 15 cm short cable with USB protection cap
- Additional USB extension cable provided
- 4 LED indicators (Num, scroll, caps lock, clean mode)
The CleanType® Easy Basic is a high-quality plastic keyboard that is easy to clean thanks to its water and dust resistance, making it ideal for use in hygiene-sensitive areas such as hospitals, clinics, medical practices, laboratories, or pharmaceutical companies. The CleanType® Easy Basic keyboard can be customized by having an individualized company logo lasered on it, integrating it perfectly into a current corporate identity. In addition, this keyboard can be equipped with any existing country layout.
Multiple cleaning methods are possible for this keyboard series. For dry cleaning, use a cleaning brush to remove dust. For wet cleaning, clean the keyboard under running warm water (max. 60 °C) with a cleaning brush (if required with commercially available cleaning agents).