Bags, stand-up, recyclable, PE/EVOH-PE
: Flexico
Clear bags, zip seal, food contact.
- Oxygen barrier and protection against humidity
- Ideal for greasy or dry products
- Suitable for food contact
- Transparency and zip seal with sound and tactile feel
- Resealable, reusable, 100% recyclable
- Upright position for shelf display
The PE/EVOH-PE bag is a 100% recyclable mono-material complex film. The presence of a 5 µm EVOH layer provides excellent barrier protection against oxygen, carbon dioxide and retains aromas, without impacting the recyclability of the bag. The layers of polyethylene surrounding the EVOH also provide the bag with a humidity protection. Your products are protected from external contamination and their properties are preserved.
: Suitable for food contact. Certificate available on request.
Frequently Bought Together