Bags, zip seal, Minigrip® bio-based
: Flexico
Clear bio-based PE bags, thickness 50 or 60 µm, food safe.
- Innovative, eco-designed and sustainable bag
- Food safe
- Totally transparent
- Hanging and/or venting hole
- Resealable, reusable, 100% recyclable
Polyethylene LDPE from renewable natural resource (sugar cane). The bag is resealable, reusable and 100% recyclable (mono-material). The Minigrip® bio-based bag provides an excellent mechanical resistance and high temperature resistance (from –40 up to +60 °C).
: Food safe certificate available on request.
Frequently Bought Together