Specifications
- Pk:2.304
- Application:
- Method / Format:Magnetic beads
- Prep size:Scalable
- Refine target molecule:Viral RNA
- Sample type:Swabs
- Target molecule:Copurification of DNA & RNA
- Description:Mag-Bind® viral DNA/RNA Xpress kit, CE IVD
- Environmentally Preferable:
- No. of reactions:24×96 preparations
Specifications
About this item
Mag-Bind® Viral DNA/RNA Xpress Kit CE IVD follows a magnetic bead-based approach for the rapid and reliable isolation of viral DNA and RNA from nasopharyngeal (NP) swab specimens that are dry or in viral transport media (VTM), from saliva and other sample sources.
Mag-Bind® Viral DNA/RNA Xpress Kit CE IVD follows a magnetic bead-based approach for the rapid and reliable isolation of viral DNA and RNA from nasopharyngeal (NP) swab specimens that are dry or in viral transport media (VTM), from saliva and other sample sources. The extraction methodology is easily adaptable to various automated systems and can also be scaled up or down depending on the amount of starting sample amount used. The kit utilizes the proven Mag-Bind® technology that enables the purification of high-quality nucleic acids that are free of proteins, nucleases, and other impurities. The purified nucleic acids are ready for direct use in downstream applications such as qPCR, RT-qPCR, and more.