Masterflex® Sanitary Clamps, Nylon, Avantor®
Supplier: Avantor Fluid Handling
Certificates
About this item
Compatible with metal and plastic Tri-Clamp® fittings.
- Excellent chemical resistance
- Rated to 150 psi
- Sterilizable via Autoclave and Gamma Irradiation
- Features anti-tamper zip-tie lock out
- Manufactured and packaged in cleanroom
Constructed of nylon, these Tri-Clamp® sanitary clamps are perfect for single-use transfer lines in biopharma markets, plus a wide range of fluid transfer systems. These clamps ideal for securing your fluid transfer system without damaging the process lines.
- Environmentally Preferable:
Specifications
- Sanitary clamp size (port 1):
- 2"
- 3"
- 3/4"
- 1" to 1-1/2"
- Connection type:
- Sanitary
- Sanitary
- Sanitary
- Sanitary
- Sterilisation:
- Autoclavable, gamma irradiation
- Autoclavable, gamma irradiation
- Autoclavable, gamma irradiation
- Autoclavable, gamma irradiation
- Warranty:
- Manufacturer defect
- Manufacturer defect
- Manufacturer defect
- Manufacturer defect