Rutaecarpine
ACRO465982500
:
- Pack type:Glass bottle
- Pk:250 mg
- Seq:2590
- Environmentally Preferable:
- Appearance:White to beige powder
- Infrared spectrum:Conforms
- Melting point:259°C to 261°C
- Loss on drying:≤ 0.5%
- HPLC:≥ 97.5%
: 8,13-Dihydroindolo[2',3':3,4]pyrido[2,1-b]quinazolin-5(7H)-one, Rutecarpine, 7,8-Dihydroindolo[2',3':3,4]pyrido[2,1-b]quinazolin-5(13H)-one
:
:287.32 g/mol
:Refrigerator
:84-26-4
:C₁₈H₁₃N₃O
:MFCD00210551
:259°C…260°C