Plastic Pallet Boxes
Supplier: MULTIROIR
Certificates
About this item
These polypropylene pallet boxes are ideal for storage, transportation and preparation of goods and/or foodstuffs. Its material gives them exceptional properties in terms of chemical resistance, shock resistance and temperature variations.
- Made of polypropylene for high impact resistance
- High load capacity and light weight
- Resistant to thermal and weather shocks
- Colour: Grey
Suitable for export to all destinations.
Lids for plastic pallet boxes
Product Information
Frequently Bought Together