Safety goggles, GoggleGear™ 6000 series
: 3M
GoggleGear™ safety goggles 6000 series features a large profile with indirect ventilation for air circulation with Scotchgard™ anti-fog and anti-scratch coating.
- Large profile safety goggles provide enough space to fit over most prescription glasses
- Indirect airflow to help against fogging
- Premium spherical curved lens design for optimal horizontal and vertical field of vision
- Scotchgard™ anti-fog coating provides superior anti-fog and anti-scratch properties
- Pivoting ratchet for convenient personal adjustment
- 2C-1.2 3M 1 BT K N lens marking
- Wide flexible nose area to fit well with half-mask respirators
These large profile safety goggles feature indirect ventilation to allow air to circulate and help reduce fogging. The polycarbonate lenses feature a Scotchgard™ anti-fog and anti-scratch coating for superior anti-fog protection and increased durability that resists fogging for longer than traditional anti-fog coatings, even after up to 25 washings with water. Available in a range of shroud colours.
Blue goggles with neoprene headband can be sterilised up to 40x by autoclaving at 121 °C.
Frequently Bought Together