Order Entry
United Kingdom
ContactUsLinkComponent
Masterflex® Adapter Fittings, Female Luer to Threaded, Straight, Avantor®
undefined
Masterflex® Adapter Fittings, Female Luer to Threaded, Straight, Avantor®
  • Model:
  • 45508-70
  • 45508-76
  • 45508-66
  • 45501-66
  • 45508-60
  • 30800-00
  • 45501-60
  • Connection type (port 1):
  • Female luer
  • Female luer
  • Female luer
  • Female luer
  • Female luer
  • Female luer
  • Female luer
  • Connection type (port 2):
  • 10-32 special tapered thread
  • 1/4-28 UNF bottom sealing thread
  • 1/4-28 UNF thread
  • 10-32 UNF thread
  • 1/8-27-NPT thread
  • No. of ports:
  • 2
  • 2
  • 2
  • 2
  • 2
  • 2
  • 2
  • Warranty:
  • 3 Months
  • 3 Months
  • 3 Months
Frequently Bought Together