Hayward® True Union Solenoid Valves
Instant on/off valve.
- Ideal for process systems requiring an instant on/off actuated valve
- Lower coil operating temperatue result in valves that are rated for 100% continuous duty
- Built-in 1/2" conduit connection
- Normally closed
- Includes both socket and NPT(F) conections
Choose between PVC and CPVC valve materials for a wide range of applications.
- Environmentally Preferable:
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.