Order Entry
United Kingdom
ContactUsLinkComponent
Moulded disposable mask, FFP3, JSP LTD
Moulded disposable mask, FFP3, JSP LTD
Catalog # 111-2586
Supplier:  JSP
undefined
Moulded disposable mask, FFP3, JSP LTD
Catalog # 111-2586
Supplier:  JSP
Specifications
  • Description:
    Moulded disposable mask, valved
  • Environmentally Preferable:
  • Pk:
    10
  • With valve:
    +
  • Class:
    FFP3
Specifications
About this item
Moulded disposable respirator with comfort foam surround, adjustable nose clip, four-point adjustable harness, and exhalation valve.
  • Exhalation valve
  • Foam padded nose bridge
  • 4-point adjustable straps
  • 99% minimum filtration efficiency
A mouldable nose bridge and 360° foam area around the inside provide comfort and a secure fit. Metal-free, 4-point adjustable straps create a firm and adaptable fit.
FFP3 rating provides 99% minimum filtration efficiency.
Certifications: Conforms to EN 149.