Tyvek® 1422A autoclavable wrappers
Improve the microbial barrier and cleanliness of your sterilisation wrap.
- Manufactured and packaged in a controlled cleanroom environment
- Autoclavable at 121 to 125 °C
- Tear and puncture resistant, hydrophobic and lint-free Tyvek® 1422A
Drapeable Tyvek® 1422A Wrappers are an improvement on cellulose or SMS based wrapping materials for steam sterilisation. Eliminate the wasted time and expense of cutting larger square sheets of sterilisation wrap to the desired sizes. Round shaped wrappers are an ideal way to cover round openings. Clean cut round and rectangular/square sheets are delivered in labelled, sealed poly bags to ensure cleanliness. Simply secure in place with autoclavable elastic bands.
: Certificates of Conformance sent with each shipment.
: Note: Dimensions shown are W×L.
Frequently Bought Together