Order Entry
United Kingdom
ContactUsLinkComponent
 
HyClone™ Fetal Bovine Serum Characterized (Australia), HyClone Products (Cytiva)
 
undefined
HyClone™ Fetal Bovine Serum Characterized (Australia), HyClone Products (Cytiva)

 

  • Catalog No:
  • SH30084.03IR25
  • SH30084.03IR30
  • Origin:
  • AU
  • AU
  • Endotoxin:
  • ≤25 EU/ml
  • ≤25 EU/ml
  • Haemoglobin:
  • ≤25 mg/dl
  • ≤25 mg/dl
  • Storage temperature:
  • Sera should be stored at −10 °C or lower. Once thawed, sera should be stored at 2...8 °C.
  • Sera should be stored at −10 °C or lower. Once thawed, sera should be stored at 2...8 °C.

 

Frequently Bought Together