J&W DB-1 GC columns, megabore
: Agilent
DB-1 is a reliable, cross-linked and non-polar column with low bleed and a high temperature limit, delivering superb performance for general purpose applications.
- 100% Dimethylpolysiloxane
- Non-polar and low bleed with a high temperature limit
- Wide range of applications
- Bonded, cross-linked and solvent rinsable
- Equivalent to USP Phase G2
Frequently Bought Together