Electron capture detectors (ECD)
: Agilent
The micro ECD has a detection zone volume 10 times smaller than any other ECD with replicable inlet liner to decreasing column contamination of the cell.
- Replaceable liner
The replaceable liner serves as a physical stop for the column, ensuring reproducible column installation and decreasing column contamination of the cell.
Frequently Bought Together