Order Entry
United Kingdom
ContactUsLinkComponent
DECON-Clean® cleaning agents
DECON-Clean® cleaning agents
  VELTDC-01-E
undefined
DECON-Clean® cleaning agents
  VELTDC-01-E
  • Description:
    DECON-Clean® Cleaning agent, concentrate, Plastic container, 3800 ml, non sterile
  • Seq:
    03
  • Environmentally Preferable:
  • Pk:
    4
Ideal for washable, nonporous surfaces such as walls, ceilings, floors, and stainless steel items, this cleaning agent is designed to remove residues left behind by disinfecting agents
DECON-Clean® is filtered at 0.2µm, gamma irradiated, and double-bagged. SimpleMix® one-step residue remover dilutes to 1:128 in Water for Injection. Sterile options are USP tested and delivered with a lot-specific certificate of irradiation and certificate of analysis. Traceable sterile cleaning agent comes with an LAL testing analysis and sterility report.