Ethyl 2,4,5-trifluorobenzoylacetate
Supplier: Apollo Scientific
Certificates
About this item
Synonyms: (2,4,5-Trifluorobenzoyl)acetic acid ethyl ester, 3-Oxo-3-(2,4,5-trifluorophenyl)propanoic acid ethyl ester, Ethyl (2,4,5-Trifluorobenzoyl)acetate, Ethyl 2,4,5-trifluorobenzoylacetate, Ethyl 3-Oxo-3-(2,4,5-trifluorophenyl)propanoate
CAS Number:98349-24-7
:
Formula:C₁₁H₉F₃O₃
MDL Number:MFCD00792431
Boiling Pt:92 °C/0.08mmHg
Molecular Weight:246.19 g/mol
Melting Pt:63…64 °C
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together