(4-Chloro-2-fluorophenyl)acetic acid 97%
Supplier: Apollo Scientific
Certificates
About this item
Synonyms: 4-Chloro-2-fluorophenylacetic acid
CAS Number:194240-75-0
:
Formula:(C₆H₃ClF)CH₂COOH
MDL Number:MFCD03095402
Molecular Weight:188.59 g/mol
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together