Order Entry
United Kingdom
ContactUsLinkComponent
4-Bromo-2,6-difluorophenylacetic acid
4-Bromo-2,6-difluorophenylacetic acid
Catalog # APOSPC8245-5G
Supplier:  Apollo Scientific
CAS Number:  
4-Bromo-2,6-difluorophenylacetic acid
Catalog # APOSPC8245-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173
  • Environmentally Preferable:

Specifications

About this item

Synonyms: 2-(4-Bromo-2,6-difluorophenyl)acetic acid
 :
 :251.027 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:537033-54-8
Formula:C₈H₅BrF₂O₂
MDL Number:MFCD03094466
Melting Pt:150…152 °C
Density:1.77 g/cm³ (25 °C)