2,6-Difluoro-3-ethoxyphenylboronic acid 98%
Supplier: Apollo Scientific
Certificates
About this item
Synonyms: 3-Ethoxy-2,6-difluorophenylboronic acid, 3-Ethoxy-2,6-difluorobenzeneboronic acid
CAS Number:849062-00-6
:
Formula:C₈H₉BF₂O₃
MDL Number:MFCD07369735
Molecular Weight:201.96 g/mol
Melting Pt:76…77 °C
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together