3'-Fluoro-2'-hydroxyacetophenone 98%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:699-92-3
:
Formula:C₈H₇FO₂
MDL Number:MFCD09055140
Molecular Weight:154.14 g/mol
Melting Pt:72…76 °C
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together