Order Entry
United Kingdom
ContactUsLinkComponent
6-Fluoro-1H-benzimidazole-2-thiol 95+%
6-Fluoro-1H-benzimidazole-2-thiol 95+%
Catalog # APOSPC201265-5G
Supplier:  Apollo Scientific
CAS Number:  
6-Fluoro-1H-benzimidazole-2-thiol 95+%
Catalog # APOSPC201265-5G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    5 g
  • Seq:
    3173
  • Environmentally Preferable:

Specifications

About this item

Synonyms: 5-Fluoro-1H-benzo[d]imidazole-2(3H)-thione
 :
 :168.19 g/mol
Storage Temperature:Ambient
CAS Number:583-42-6
Formula:C₇H₅FN₂S
MDL Number:MFCD07432806