Order Entry
United Kingdom
ContactUsLinkComponent
Potassium(2-naphthalene)trifluoroborate 96%
Potassium(2-naphthalene)trifluoroborate 96%
Catalog # APOSPC1943-500MG
Supplier:  Apollo Scientific
CAS Number:  
Potassium(2-naphthalene)trifluoroborate 96%
Catalog # APOSPC1943-500MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    500 mg
  • Seq:
    2725
  • Environmentally Preferable:

Specifications

About this item

Synonyms: Potassium 2-Naphthalenetrifluoroborate, (2-Naphthalene)trifluoroboric acid potassium salt, Potassium trifluoro(naphthalen-2-yl)borate, Potassium trifluoro(2-naphthalenyl)borate
 :
 :234.07 g/mol
 :GHS07
Storage Temperature:Ambient
CAS Number:668984-08-5
Formula:C₁₀H₇BF₃K
MDL Number:MFCD06659941
Melting Pt:>300 °C