3,5-Difluoro-2-methoxyphenylboronic acid 96%
Supplier: Apollo Scientific
Certificates
About this item
Synonyms: (3,5-Difluoro-2-methoxyphenyl)boronic acid, 3,5-Difluoro-2-methoxybenzeneboronic acid
CAS Number:737000-76-9
:
Formula:C₇H₇BF₂O₃
MDL Number:MFCD03095353
Molecular Weight:187.94 g/mol
Melting Pt:112…115 °C
:GHS07
Storage Temperature:Freezer
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together