3-(Trifluoromethyl)-p-anisidine 98%
Supplier: Apollo Scientific
Certificates
About this item
Synonyms: 3-(Trifluoromethyl)-4-anisidine, 5-Amino-2-methoxybenzotrifluoride, 4-Methoxy-3-(trifluoromethyl)aniline, 3-(Trifluoromethyl)-4-methoxyaniline, 4-Amino-2-(trifluoromethyl)anisole
CAS Number:393-15-7
:
Formula:C₈H₈F₃NO
MDL Number:MFCD00043460
Boiling Pt:112 °C/18mmHg
Molecular Weight:191.15 g/mol
Melting Pt:58…59 °C
Storage Temperature:Ambient
UN:2811
ADR:6.1 III
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together