4-(Methylsulphonyl)benzenesulphonyl chloride 95+%
Catalog # APOSOR9678-250MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:250 mg
- Seq:2590
- Environmentally Preferable:
Specifications
About this item
Synonyms: 4-(Methylsulphonyl)benzene-1-sulphonyl chloride
:
:254.699 g/mol
UN:3261
:GHS05
Storage Temperature:Ambient
CAS Number:82964-91-8
Formula:C₇H₇ClO₄S₂
MDL Number:MFCD00216486
Melting Pt:165…169 °C
ADR:8 III