3-Nitropyrrole 95%
Supplier: Apollo Scientific
Certificates
About this item
Synonyms: 3-Nitro-1H-pyrrole
CAS Number:5930-94-9
:
Formula:C₄H₄N₂O₂
MDL Number:MFCD00059714
Molecular Weight:112.09 g/mol
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together