2-Methylthiazole-5-methanol 95%
Supplier: Apollo Scientific
Certificates
About this item
Synonyms: (2-Methylthiazol-5-yl)methanol
CAS Number:56012-38-5
:
Formula:C₅H₇NOS
MDL Number:MFCD12912988
Molecular Weight:129.18 g/mol
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together