Sodium ethylxanthate 95%
Supplier: Apollo Scientific
Certificates
About this item
Synonyms: Sodium Xanthogenate, Ethylxanthic Acid Sodium Salt, Sodium O-Ethyl Dithiocarbonate
CAS Number:140-90-9
:
Formula:C₃H₆OS₂
MDL Number:MFCD00068314
Molecular Weight:144.18 g/mol
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together