Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
£0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
United Kingdom
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-Amino-1,2,5-oxadiazole-3-carboxylic acid
4-Amino-1,2,5-oxadiazole-3-carboxylic acid 96%
4-Amino-1,2,5-oxadiazole-3-carboxylic acid 96%
4-Amino-1,2,5-oxadiazole-3-carboxylic acid 96%
APOSOR912026-25G
:
Apollo Scientific
:
4-Amino-1,2,5-oxadiazole-3-carboxylic acid 96%
APOSOR912026-25G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
25 g
Seq:
3308
Environmentally Preferable:
:
:
129.0742 g/mol
:
GHS07
:
Ambient
:
78350-50-2
:
C₃H₃N₃O₃
:
MFCD00235172