Isopropylhydrazine hydrochloride
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:16726-41-3
:
Formula:C₃H₁₁ClN₂
MDL Number:MFCD01746887
Molecular Weight:110.5858 g/mol
Melting Pt:115…118 °C
:GHS07
Storage Temperature:Refrigerator
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together