3-Amino-4-methylphenol
Supplier: Apollo Scientific
Certificates
About this item
Synonyms: 3-Amino-p-cresol, 5-Hydroxy-2-methylaniline
CAS Number:2836-00-2
:
Formula:C₇H₉NO
MDL Number:MFCD00075083
Molecular Weight:123.16 g/mol
Melting Pt:156…157 °C
EINECS:220-622-8
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Frequently Bought Together